Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant L-lactate dehydrogenase (LDH)
Mouse recombinant L-lactate dehydrogenase (LDH)
Catalog # PRSI92-414
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant L-lactate dehydrogenase (LDH)
Catalog # PRSI92-414
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    L-Lactate dehydrogenase (LDH)
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    37,5 kDa
  • Sequence:
    Met1-Phe332
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Mouse Lactate dehydrogenase A (LDHA) is a member of the LDH/MDH superfamily and LDH family. LDHA catalyses the inter-conversion of pyruvate and L-lactate with concomitant inter-conversion of NADH and NAD+. LDHA is found in most somatic tissues, though predominantly in muscle tissue and tumours. It has also been shown that LDHA plays an important role in the development, invasion and metastasis of malignancies. Mutations in LDHA have been linked to exertional myoglobinuria.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.