Order Entry
Switzerland
ContactUsLinkComponent
 
Mouse recombinant plasma glutamate carboxypeptidase
Catalog # PRSI92-411
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant plasma glutamate carboxypeptidase
Catalog # PRSI92-411
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Plasma glutamate carboxypeptidase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    50,9 kDa
  • Sequence:
    Lys19-Ser470
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Carboxypeptidase Q (Cpq) is a member of the peptidase M28 family. PGCP is involved in a number of fundamental biological processes such as the hydrolysis of circulating peptides, catalyzing the hydrolysis of dipeptides with unsubstituted terminals into amino acids. Carboxypeptidase may play an important role in the liberation of thyroxine hormone from its thyroglobulin (Tg) precursor. The monomeric form is inactive while the homodimer is active.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.