Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Thymidylate kinase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:24.9 kD
- Sequence:Met1-Lys212
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
Thymidylate kinase (DTYMK) is a ubiquitous enzyme of about 25 kD which belongs to thymidylate kinase family. DTYMK is important in the dTTP synthesis pathway for DNA synthesis. It participated in the pyrimidine metabolism pathway and dTTP biosynthesis pathway. DTYMK catalyses the conversion of dTMP to dTDP and catalyses the phosphorylation of thymidine 5'-monophosphate (dTMP) to form thymidine 5'-diphosphate (dTDP) in the presence of ATP and magnesium. Structural and functional analyses suggest that the cDNA codes for authentic human dTMP kinase. The mRNA levels and enzyme activities corresponded to cell cycle progression and cell growth stages.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.