Human recombinant phosphoglycerate kinase 1 (from cells)
Catalog # PRSI92-357
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Phosphoglycerate kinase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:45.5 kD
- Sequence:Ser2-Ile417
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl, 20% glycerol,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
PGK1 is called phosphoglycerate kinase that involved in a critical energy-producing process known as glycolysis. Phosphoglycerate kinase helps carry out a chemical reaction that converts a molecule called 1,3-diphosphoglycerate, which is produced during the breakdown of glucose, to another molecule called 3-phosphoglycerate during glycolysis. PGK1 The encoded protein may also act as a cofactor for polymerase alpha.. The protein has been identified as a moonlighting protein based on its ability to perform mechanistically distinct functions.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.