Human recombinant leukotriene A4 hydrolase (from cells)
Catalog # PRSI92-331
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Leukotriene A4 Hydrolase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:70.3 kD
- Sequence:Met1-Asp611
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM MES,2 mM EDTA, 20% glycerol,pH5.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
LTA4H, which is short for Leukotriene A-4 hydrolase, is a 611 aa. protein. It belongs to the peptidase M1 family, and exists in cytoplasm. This protein has at least 4 isforms produced by alternative splicing, and two of them are expressed in monocytes, lymphocytes, neutrophils, reticulocytes, platelets and fibroblasts. LTA4H involves in lipid metabolism and leukotriene B4 biosynthesis. It is a epoxide hydrolase that catalyses the final step in the biosynthesis of the proinflammatory mediator leukotriene B4. It also has aminopeptidase activity.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.