Order Entry
Switzerland
ContactUsLinkComponent
E. coli recombinant beta-glucuronidase (from E. coli)
E. coli recombinant beta-glucuronidase (from E. coli)
Catalog # PRSI92-300
Supplier:  ProSci Inc.
CAS Number:  
E. coli recombinant beta-glucuronidase (from E. coli)
Catalog # PRSI92-300
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    E. coli
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    beta-Glucuronidase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    69.9 kD
  • Sequence:
    Met1-Gln603
  • Formulation:
    Supplied as a 0.2 µm filtered solution of PBS,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Beta-D-glucuronidase from E. coli is a  highly specific enzyme in hydrolysing glucuronides. Beta-D-glucuronidase (GLUase) activity can be measured as the rate of production of fluorescent methylumbelliferone (MU), resulting from the hydrolysis of the substrate 4-methylumbelliferyl- beta -d-glucuronide (MUGLU), which is an effective and rapid method for detection and verification of E. coli in food, water, and environmental samples. High purity recombinant beta-Glucuronidase is used in research, biochemical enzyme assays and in vitro diagnostic analysis, detecting a wide range of drugs such as opioids, benzodiasepines, steroids, cannabinoids, and others.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.