Human recombinant Methenyl-THF synthetase (from E. coli)
Catalog # PRSI92-280
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Methenyl-THF synthetase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:24.3 kD
- Sequence:Met1-Ala203
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris,200 mM Nacl,1 mM DTT,50% Glycerol,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
5-formyltetrahydrofolate cyclo-ligase (MTHFS) belongs to the 5-formyltetrahydrofolate cyclo-ligase family. It is an enzyme that catalyses the conversion of 5-formyltetrahydrofolate to 5,10-methenyltetrahydrofolate, contributes to tetrahydrofolate metabolism. MTHFS helps regulate carbon flow through the folate-dependent one-carbon metabolic network that supplies carbon for the biosynthesis of purines, thymidine and amino acids. An increased activity of the encoded protein can result in an increased folate turnover rate and folate depletion.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.