Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant ATPase SWSAP1 (from E. coli)
Human recombinant ATPase SWSAP1 (from E. coli)
Catalog # PRSI92-241
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ATPase SWSAP1 (from E. coli)
Catalog # PRSI92-241
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    ATPase SWSAP1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.7 kD
  • Sequence:
    Met1-Pro229
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

SWSAP1 is a nucleus ATPase protein, interacts with ZSWIM7 and forms a functional complex. The complexs involved in homologous recombination repair and stabilises each other. SWS1AP1 also interacts with RAD51, RAD51B, RAD51C, RAD51D and XRCC3. It involves in homologous recombination repair. ATPase is preferentially stimulated by single-stranded DNA and is involved in homologous recombination repair (HRR). SWSAP1 has a DNA-binding activity which is independent of its ATPase activity.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.