Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant CaM kinase ID (from E. coli)
Human recombinant CaM kinase ID (from E. coli)
Catalog # PRSI92-220
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CaM kinase ID (from E. coli)
Catalog # PRSI92-220
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-GST tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    CaM kinase ID
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    69.2 kD
  • Sequence:
    Met1-Lys385
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB,150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

CAMK1D is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It contains a protein kinase domain. CAMK1D expressed in polymorphonuclear leukocytes and may be part of the chemokine signal transduction pathway that regulates granulocyte function. It may also be involved in modulation of neuronal apoptosis. It activates CREB-dependent gene transcription, regulates calcium-mediated granulocyte function and respiratory burst and promotes basal dendritic growth of hippocampal neurons.

Fusion-Tag: N-GST tag

This recombinant protein can be used for biological assays. For research use only.