Human recombinant Peptidase D (from E. coli)
Catalog # PRSI92-214
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Peptidase D
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:54.5 kD
- Sequence:Ala2-Lys493
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB,150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
PEPD belongs to the peptidase M24B family of Eukaryotic-type prolidase subfamily. PEPD is a cytosolic dipeptidase that hydrolyses dipeptides with proline or hydroxyproline at the carboxy terminus. It is important in collagen metabolism because of the high levels of imino acids. Defects in PEPD are a cause of prolidase deficiency which is an autosomal recessive disorder associated with iminodipeptiduria.
This recombinant protein can be used for biological assays. For research use only.