Human recombinant sialic acid synthase (from E. coli)
Catalog # PRSI92-137
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Sialic Acid Synthase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:42.4 kD
- Sequence:Met1-Ser359
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris, 100 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Sialic Acid Synthase (NANS) is an enzyme that contains one AFP-like domain. NANS is ubiquitous and plays a role in the biosynthetic pathways of sialic acids. NANS produces N-acetylneuraminic acid (Neu5Ac) and 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN). It also can use N-acetylmannosamine 6-phosphate and mannose 6-phosphate as substrates to generate phosphorylated forms of Neu5Ac and KDN, respectively.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.