Technische Daten
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Caspase 10
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:30.1 kD
- Sequence:Val220-Ile480
- Formulation:Supplied as a 0.2 µm filtered solution of 25 mM HEPES, 10 mM DTT, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Technische Daten
Über diesen Artikel
Caspase-10 (CASP10) is a 521 amino acid protein member of the Cysteine-Aspartic Acid Protease (Caspase) family. CASP10 contains two DED (Death Effector) domains and is detectable in most tissues. CASP10 cleavage by Granzyme B and autocatalytic activity generate the two active subunits: Caspase-10 subunit p23/17, Caspase-10 subunit p12. Caspases are a family of cytosolic aspartate-specific cysteine proteases involved in the execution-phase of cell apoptosis, the initiation and execution. Human caspases can be subdivided into three functional groups: cytokine activation (caspase-1, -4, -5, and -13), apoptosis initiation (caspase-2, -8, -9, -and -10), and apoptosis execution (caspase-3, -6, and -7). CASP10 cleaves and activates caspases 3 and 7, but itself is processed by caspase 8. Defects in CASP10 are associated with apoptosis defects seen in type II autoimmune lymphoproliferative syndrome.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.