Human recombinant Lysophospholipase II (from E. coli)
PRSI92-096
:
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Lysophospholipase II
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:25.8 kD
- Sequence:Met1-Val231
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris, 1 mM DTT, 0.1 M NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Acyl-Protein Thioesterase 2 is a cytoplasmic protein that belongs to the AB Hydrolase 2 family. LYPLA2 is a lysophospholipase and hydrolyses fatty acids from S-acylated cysteine residues in proteins such as trimeric G alpha proteins or HRAS. LYPLA2 has hydrolase activity that converts Palmitoyl-protein to palmitate and protein. LYPLA2 regulates the multifunctional lysophospholipids by acting on biological membranes. LYPLA2 participates in Glycerophospholipid metabolism pathway, fatty acid metabolic process and lipid metabolic process.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.