Human recombinant ribulose-phosphate 3-epimerase (from E. coli)
PRSI92-090
:
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Ribulose-Phosphate 3-Epimerase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:25.9 kD
- Sequence:Met1-Arg228
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 6.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Ribulose-Phosphate 3-Epimerase (RPE) is a member of the Ribulose-Phosphate 3-Epimerase family. RPE exists as a homodimer and catalyses the reversible epimerisation of D-ribulose 5-phosphate to D-xylulose 5-phosphate. RPE binds one divalent metal cation per subunit and contains tightly bound Fe2+ when produced in E. coli, but the physiological cofactor may be Co2+, Mn2+ or Zn2+. It has been shown that RPE participates in 3 metabolic pathways: pentose phosphate pathway, pentose and glucuronate interconversions, and carbon fixation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.