Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant PPIase D (from E. coli)
Human recombinant PPIase D (from E. coli)
Catalog # PRSI92-055
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant PPIase D (from E. coli)
Catalog # PRSI92-055
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N, C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    PPIase D
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    43.9 kD
  • Sequence:
    Met1-Ala370
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Used for biological assays

Specifications

About this item

Peptidyl-Prolyl Cis-Trans Isomerase D (PPID) belongs to the cyclophilin-type PPIase family and PPIase D subfamily. PPID is widely expressed and it contains one PPIase cyclophilin-type domain and three TPR repeats. PPID catalyses the cis-trans isomerisation of proline imidic peptide bonds in oligopeptides and accelerates the folding of proteins. PPID can bind to the immunosuppressant cyclosporine A and is known that its overexpression suppresses the apoptosis in cancer cells.

Fusion-Tag: N, C-6 His tag

This recombinant protein can be used for biological assays. For research use only.