Human recombinant Selenocysteine lyase (from E. coli)
Catalog # PRSI92-018
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Selenocysteine Lyase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:50.34 kD
- Sequence:Met1-Ala445
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Selenocysteine Lyase belongs to the class-V pyridoxal-phosphate-dependent aminotransferase family. Selenocysteine Lyase exists as a homodimer in the cytosol. In the brain, Selenocysteine Lyase is as an enzyme that putatively salvages Sec and recycles the selenium for selenoprotein translation. Selenocysteine Lyase catalyses the decomposition of L-selenocysteine to L-alanine and elemental selenium. Selenocysteine Lyase can be up-regulated In acute glomerulonephritis, it can also be regulated by JUN/AP-1.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.