Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant IPP Isomerase 2 (from E. coli)
Human recombinant IPP Isomerase 2 (from E. coli)
Catalog # PRSI92-015
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IPP Isomerase 2 (from E. coli)
Catalog # PRSI92-015
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    IPP Isomerase 2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28.9 kD
  • Sequence:
    Met1-Val227
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 1 mM DTT, 0.1 mM PMSF, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Isopentenyl Pyrophosphate Isomerase 2 (IDI2) belongs to the IPP isomerase type 1 family. Both isozymes, IDI1 and IDI2 are localised to the peroxisome by a PTS1-dependent pathway. IDI2 is expressed in skeletal muscle, which contains one nudix hydrolase domain. IDI2 binds one magnesium per subunit. IDI2 catalyses the 1,3-allylic rearrangement of the homoallylic substrate isopentenyl (IPP) to its highly electrophilic allylic isomer, dimethylallyl diphosphate (DMAPP). It is reported that IDI2 is regulated independently from IDI1, by a mechanism that may involve PPAR- alpha.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.