Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Argininosuccinate Synthase (from E. coli)
Human recombinant Argininosuccinate Synthase (from E. coli)
Catalog # PRSI91-986
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Argininosuccinate Synthase (from E. coli)
Catalog # PRSI91-986
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Argininosuccinate Synthase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    42.8 kD
  • Sequence:
    Met1-Lys412
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, 50 mM Imidazole, 1 mM DTT, 40% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Argininosuccinate Synthase (ASS1) is an urea cycle enzyme with a tetrameric structure composed of identical subunits. ASS1 is involved in the synthesis of arginine and catalyses that condensation of citrulline and aspartate to argininosuccinate using ATP. ASS1 is important to the urea cycle as it catalyses the important second last step in the arginine biosynthetic pathway. ASS1 mainly expressed in periportal hepatocytes, but also in most other body tissues. A deficiency of ASS1 causes citrullinemia (CTLN1), an autosomal recessive disease which is characterised by severe vomiting spells and mental retardation.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.