Human Recombinant Glutamine Synthetase (from E. coli)
Catalog # PRSI91-970
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Glutamine Synthetase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:43.13 kD
- Sequence:Thr2-Asn373
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 200 mM NaCl, 50 mM Imidazole, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Glutamine Synthetase reglutes intracellular concentration of glutamate. Glutamine Synthetase catalyses the synthesis of glutamine from glutamate and ammonia. Glutamine is an important source of energy and that takes part in cell prolifetation, inhibition of apoptosis, and cell signaling. Glutamine Synthetase is expressed during early fetal stages, and has a role in maintaining body PH by removing ammonia from circulation. Mutations in the GLUL gene are related to congenital glutamine deficiency.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.