Human recombinant N-acetylgalactosaminyltransferase 7 (from cells)
Catalog # PRSI91-921
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:N-acetylgalactosaminyltransferase 7
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:73.2 kD
- Sequence:Pro30-Val657
- Formulation:Supplied as a 0.2 µm filtered solution of 50 mM TrisHCl,10 mM reduced Glutathione,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
N-acetylgalactosaminyltransferase 7(GALNT7) is expressed in uterus, retina, kidney, small intestine, omentum, stomach and CNS.It belongs to the glycosyltransferase 2 family and galNAc-T subfamily. The enzyme encoded by this gene controls the initiation step of mucin-type O-linked protein glycosylation and transfer of N-acetylgalactosamine to serine and threonine amino acid residues. This enzyme is a type II transmembrane protein and shares common sequence motifs with other family members. This protein may function as a follow-up enzyme in the initiation step of O-glycosylation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.