Human recombinant Creatine Kinase (from cells)
Catalog # PRSI91-873
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Creatine Kinase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:44.1 kD
- Sequence:Met1-Lys381
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Creatine kinase M-type is also known as Creatine kinase M chain,M-CK. It is a protein that in humans is encoded by the CKM gene. It belongs to the ATP:guanido phosphotransferase family,containing 1 phosphagen kinase C-terminal domain and 1 phosphagen kinase N-terminal domain. Creatine kinase M-type can reversibly catalyses the transfer of phosphate between ATP and various phosphogens. It plays a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.