Human recombinant ketohexokinase (from cells)
Catalog # PRSI91-843
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Ketohexokinase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:33.7 kD
- Sequence:Met1-Val298
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 50nM KCl, 10% Glycerol, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Ketohexokinase, also known as Hepatic fructokinase, is a member of the carbohydrate kinase PfkB family. It exits as a homodimer and most abundant in liver, kidney, gut, spleen and pancreas. Low levels also found in adrenal, muscle, brain and eye.This enzyme catalyses conversion of fructose to fructose-1-phosphate. It is the first enzyme with a specialised pathway that catabolises dietary fructose. Defects in KHK are the cause of fructosuria.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.