Human recombinant PPIase FKBP2 (from cells)
Catalog # PRSI91-839
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:PPIase FKBP2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:14.3 kD
- Sequence:Ala22-Leu142
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Used for biological assays
Specifications
About this item
Peptidyl-prolyl cis-trans isomerase FKBP2(FKBP2 for short), also named 13 kDa FK506-binding protein, FK506-binding protein 2, Immunophilin FKBP13, Rotamase, is a endoplasmic reticulum peripheral membrane protein. It contains 1 PPIase FKBP-type domain and belongs to the FKBP-type PPIase family, FKBP2 subfamily which takes part in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP2 is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. FKBP2 functions as an ER chaperone and as a component of membrane cytoskeletal scaffolds.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.