Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Alkaline Phosphatase (from cells)
Human recombinant Alkaline Phosphatase (from cells)
  PRSI91-795
 :  
Human recombinant Alkaline Phosphatase (from cells)
  PRSI91-795
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Alkaline Phosphatase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    53.7 kD
  • Sequence:
    Ile23-Asp506
  • Formulation:
    Supplied as a 0.2 µm filtered solution of PBS, pH 7.4 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

ALPP is a membrane protein and exits as a homodimer. ALPP is expressed only in normal term placenta, endocervix and fallopian tube and also in ovarian and proximal gastrointestinal tumors. It has been shown to play a role in a number of processes including cell signaling, long-term potentiation, and cell adhesion, however, the best known and most commonly studied role is implicated in Alzheimer's research.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.