Human recombinant protein Disulfide-Isomerase A4 (from cells)
PRSI91-762
:
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Protein Disulfide-Isomerase A4
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:71.7 kD
- Sequence:Val21-Leu645
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Protein Disulfide-Isomerase A4 (PDIA4) is an endoplasmic reticulum luminal protein that belongs to the protein disulfide isomerase family. Human PDIA4 is synthesised as a 625 amino acid precursor that contains a 20 amino acid signal sequence, and a 625 amino acid mature chain, including three thioredoxin domains. PDIA4 catalyses the rearrangement of -S-S- bonds in proteins and is thought to be a deoxycytidine kinase. In addition, PDIA4 serves as a proteases protein disulfide isomerase, phospholipase or an arrangement of these.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.