Human recombinant Pancreatic Lipase-Related 2 (from cells)
PRSI91-758
:
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Pancreatic Lipase-Related 2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:51.16 kD
- Sequence:Lys18-Cys469
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM GaCl₂, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Pancreatic Lipase-Related Protein 2 (PNLIPRP2) is a secreted protein that belongs to the Lipase family of AB hydrolase superfamily. PNLIPRP2 is a lipase with broad substrate specificity that contains one PLAT domain. PNLIPRP2 is expressed mainly in pancreas. PNLIPRP2 can hydrolyse both phospholipids and galactolipids. PNLIPRP2 acts preferentially on monoglycerides, phospholipids and galactolipids. PNLIPRP2 also contributes to milk fat hydrolysis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.