Human recombinant PPIase FKBP7 (from cells)
Catalog # PRSI91-742
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:PPIase FKBP7
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:23.94 kD
- Sequence:Phe23-Val423
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM GaCl₂, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Peptidyl-Prolyl Cis-Trans Isomerase FKBP7 (FKBP7) is a member of the FKBP-type peptidyl-prolyl cis/trans isomerase (PPIase) family. FKBP7 contains two EF-hand domains and one PPIase FKBP-type domain. FKBP7 exhibits PPIase activity and function as molecular chaperones. In addition, FKBP7 accelerates the folding of proteins during protein synthesis. It has been shown that Hsp90 complex to the nucleus bind its PPIase domain to cytoplasmic dynein, the motor protein responsible for retrograde movement along microtubules.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.