Human recombinant GMP Reductase 1 (from cells)
Catalog # PRSI91-666
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:GMP Reductase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:38.5 kD
- Sequence:Met1-Ser345
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris-HCl,40% glycerol,0.15M NaCl and 1 mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
GMP Reductase 1 (GMPR) is a member of the IMPDH/GMPR family. GMPR exists as a homotetramer and catalyses the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides. GMP reductase gene expression may be regulated by MITF. At least two different alleles are known.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.