Human recombinant transaldolase (from cells)
Catalog # PRSI91-656
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Transaldolase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:38.5 kD
- Sequence:Met1-Lys337
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Transaldolase (TALDO1) belongs to the transaldolase family of Type 1 subfamily. TALDO1 is expressed selectively in oligodendrocytes of the brain. TALDO1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. TALDO1 deficiency results in telangiectases of the skin, hepatosplenomegaly and enlarged clitoris.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.