Human recombinant Carbonic Anhydrase 11 (from Cells)
PRSI91-650
:
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Carbonic Anhydrase 11
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:35.1 kD
- Sequence:His24-Arg328
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Carbonic Anhydrase-Related Protein 11 (CA11) is a secreted protein member of the alpha-carbonic anhydrase family. Carbonic Anhydrases (CAs) are a large family of zinc metalloenzymes that catalyse the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. CA11 is expressed abundantly in the brain with moderate expression also present in spinal cord and thyroid. CA11 may play a general role in the central nervous system.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.