Human recombinant beta-1,4-Galactosyltransferase 4 (from Cells)
Catalog # PRSI91-464
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:beta-1,4-Galactosyltransferase 4
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:36.77 kD
- Sequence:Gln39-Ala344
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
beta -1,4-galactosyltransferase 4 (B4GALT4) is a single-pass type II membrane protein that belongs to the Glycosyltransferase 7 family. B4GALT4 consist of the following 2 domains: N-Acetyllactosamine Synthase and beta -N-Acetylglucosaminyl-Glycolipid beta -1,4-Galactosyltransferase. B4GALT4 is highly expressed in the heart, placenta, kidney, and pancreas; it is lowly expressed in the brain, colon, lung, muscle, ovary, testis, and uterus. B4GALT4 function is responsible for the synthesis of complex-type N-linked oligosaccharides in many glycoproteins as well as the carbohydrate moieties of glycolipids.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.