Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant tryptase beta-2 (from cells)
Human recombinant tryptase beta-2 (from cells)
Catalog # PRSI91-456
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant tryptase beta-2 (from cells)
Catalog # PRSI91-456
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Tryptase beta-2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    29.64 kD
  • Sequence:
    Ala19-Pro275
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

Tryptases are Trypsin-like Serine Proteases. beta -Tryptases are the main isoenzymes in mast cells. beta tryptases form active tetramers with heparin proteoglycan. In the tetramer, the unique arrangement of the active sites facing a narrow central pore, beta -Tryptases are resistant to macromolecule protease inhibitors . When mast cells are activated, beta -Tryptases are released and participate in provoking inflammatory conditions . beta -Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic disorders.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.