Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant Chymotrypsin-like Protease (from cells)
  PRSI91-414
 :  
Human recombinant Chymotrypsin-like Protease (from cells)
  PRSI91-414
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Chymotrypsin-Like Protease
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    27.15 kD
  • Sequence:
    Cys19-Asn264
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Chymotrypsin-Like Protease CTRL-1 is a protease that belongs to the peptidase S1 family. Human CTRL-1 is synthesised as a 264 amino acid (aa) precursor that contains an 18 aa signal sequence, 15 aa activation peptide and a 231 aa mature chain. CTRL-1 Contains 1 peptidase S1 domain. It has many molecular functions, such as hydrolase, protease, and serine protease. CTRL-1 plays a role in digest and hydrolyse proteins in biological process.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.