Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant tryptase epsilon (from cells)
Catalog # PRSI91-351
Anbieter:  ProSci Inc.
CAS-Nummer:  
Human recombinant tryptase epsilon (from cells)
Catalog # PRSI91-351
Anbieter:  ProSci Inc.
CAS-Nummer:  

Technische Daten

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Tryptase epsilon
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    31.56 kD
  • Sequence:
    Ala33-Ser317
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM HAc-NaAc, 150 mM NaCl, 10% Glycerol, pH 4.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Technische Daten

Über diesen Artikel

Brain-Specific Serine Protease 4 (BSSP-4) is a serine protease that preferentially cleaves the synthetic substrate H-D-Leu-Thr-Arg-pNA compared to tosyl-Gly-Pro-Arg-pNA. BSSP-4 is expressed abundantly in the epithelial cells of the airways, including trachea, esophagus and fetal lung, but scarce in adult lung and expressed at low levels in placenta, pancreas, prostate and thyroid gland. BSSP-4 belongs to the peptidase S1 family and related to trypsin, referentially hydrolyzing substrates after arginine and lysine residues. However, BSSP-4 is less susceptible to inhibition by common trypsin inhibitors such as aprotinin, alpha1-antitrypsin and secretory leukocyte protease inhibitor. BSSP-4 efficiently converts pro-urokinase- type plasminogen activator to its mature, active form.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.