Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Arylsulfatase A (from Cells)
Human recombinant Arylsulfatase A (from Cells)
  PRSI91-281
 :  
Human recombinant Arylsulfatase A (from Cells)
  PRSI91-281
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Arylsulfatase A
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    52.9 kD
  • Sequence:
    Arg19-Ala507
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Arylsulfatase A (ARSA) is a lysosomal enzyme that breaks down Cerebroside 3-Sulfate, a major constituent of the myelin sheet, into Cerebroside and Sulfate. The ARSA deficiency results in Metachromatic Leukodystrophy (MLD), a lysosomal storage disease in the central and peripheral nervous systems with severe and progressive neurological symptoms. Recombinant Human ARSA corresponds to the ARSA mature peptide and has sulfatase activity.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.