Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant L-Xylulose Reductase (from E. coli)
Human recombinant L-Xylulose Reductase (from E. coli)
Catalog # PRSI91-257
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant L-Xylulose Reductase (from E. coli)
Catalog # PRSI91-257
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    L-Xylulose Reductase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28.1 kD
  • Sequence:
    Met1-Cys244
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 50 mM Tris, 150 mM NaCl, 1 mM DTT, 30% Glycerol, 1 mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

L-Xylulose Reductase is an enzyme that belongs to the Short-Chain Dehydrogenases/Reductases (SDR) family. L-Xylulose Reductase is responsible for the metabolism of Xylulose, converting it into Xylitol. L-Xylulose Reductase catalyses the NADPH-dependent reduction of several Pentoses, Tetroses, Trioses, alpha-Dicarbonyl compounds and L-Xylulose. L-Xylulose Reductase participates in the Uronate Cycle of Glucose metabolism. It may play a role in the water absorption and cellular osmoregulation in the proximal renal tubules by producing Xylitol, an osmolyte, thereby preventing osmolytic stress from occurring in the renal tubules.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.