Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant NAD kinase (from E. coli)
Human recombinant NAD kinase (from E. coli)
Catalog # PRSI91-250
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant NAD kinase (from E. coli)
Catalog # PRSI91-250
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    NAD Kinase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    44.4 kD
  • Sequence:
    Ser64-Gly446
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 50 mM TrisHCl, 150 mM NaCl, 1 mM DTT, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

NAD Kinase (NADK) is an enzyme that belongs to the NAD Kinase family. It is a widely expressed enzyme, but it is not detected in skeletal muscle. NADK converts Nicotinamide Adenine Dinucleotide (NAD+) into NADP+, through phosphorylating the NAD+ coenzyme. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis. The structure of the NADK from the archaean Archaeoglobus fulgidus has been determined.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.