Human recombinant Sentrin-specific protease 8 (from E. coli)
Catalog # PRSI91-152
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Sentrin-specific Protease 8
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:26.24 kD
- Sequence:Met1-Lys212
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Sentrin-Specific Protease 8 (SENP8) mediates the reversible covalent modification of proteins by NEDD8. SENP8 catalyses the full-length NEDD8 to generate its mature form and deconjugation of NEDD8 from targeted proteins such as CUL2 , CUL4A in vivo, or p53. but it does not show activity against ubiquitin or SUMO proteins.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.