Human recombinant Carbonic Anhydrase 7 (from E. coli)
Catalog # PRSI91-094
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Carbonic Anhydrase 7
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:30.72 kD
- Sequence:Met1-Ala264
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Carbonic Anhydrase 7 (CA7) is a member of the alpha-carbonic anhydrase family. Alpha-carbonic anhydrase is a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Furthermore, Alpha-carbonic anhydrase is associated with many biological processes, including calcification, respiration, bone resorption, acid-base balance and the formation of aqueous humor. CA7 is activated by histamine, L-adrenaline, L- and D-histidine, and L- and D-phenylalanine, but it is inhibited coumarins, sulfonamide derivatives such as acetazolamide (AZA) by saccharin and Foscarnet.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.