Mouse recombinant IL2 (from CHO cells)
Catalog # PRSI90-467
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Tag sequence:Fc Tag
- Storage conditions:Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Protein synonyms:Interleukin-2|Aldesleukin
- Protein/peptide name:IL2
- Purity:>98% (SDS-PAGE)
- Sequence:21-169
- Formulation:Lyophilized from 0.2um-filtered solution in PBS.
Specifications
About this item
Interleukin-2 (IL-2) is a 133 amino acid glycoprotein with one intramolecular disulfide bond and variable glycosylation. It is secreted by activated T cells and induces proliferation and maturation of activated T cells, natural killer cells, and lymphokine activated killer cells. IL-2 also stimulates proliferation of antibody-producing B cells, activates neutrophils, and induces mononuclear cells to secrete IFN-gamma and TNF-alpha and -beta. Moreover, studies have shown that IL-2 is required for activation-induced apoptosis, an important hemeostatic mechanism in the immune system, which is involved in the maintenance of peripheral tolerance to self-antigens.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.