Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant CD134 (from E. coli)
Human Recombinant CD134 (from E. coli)
Catalog # PRSI90-408
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant CD134 (from E. coli)
Catalog # PRSI90-408
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    TXGP1L|ACT35|Tumor Necrosis Factor Receptor Superfamily Member 4|OX40|TNFRSF4
  • Protein/peptide name:
    CD134
  • Purity:
    >97% (SDS-PAGE)
  • Sequence:
    29-216
  • Formulation:
    Lyophilized from a concentrated sterile solution containing 50mM Tris-HCl buffer (pH 8.0) and 500mM NaCl.

Specifications

About this item

CD134 (OX40) is a T cell activation antigen structurally belonging to a lymphocyte-specific subgroup of the nerve growth factor and tumour necrosis factor receptor superfamily, which also includes the T cell antigen CD27, B cell antigen CD40, FAS antigen and the T cell activation antigen 4-1BB. The human CD134 protein is expressed only on activated CD4+ T blasts, and its ligand has been identified as gp34. Interactions between CD134 and its ligand in vivo are necessary for the differentiation of activated B cells into highly immunoglobulin-producing cells, however not involved in other pathways of antigen-driven differentiation of B cells such as development of memory cells in the germinal centers. In addition, the CD134 and gp34 system directly mediate adhesion of activated T cells to vascular endothelial cells, and contribute to growth stimulation of the virus-infected T cells.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.