Order Entry
Switzerland
ContactUsLinkComponent
Recombinant TDO Anopheles gambiae (from E. coli)
Recombinant TDO Anopheles gambiae (from E. coli)
Catalog # PRSI90-359
Supplier:  ProSci Inc.
CAS Number:  
Recombinant TDO Anopheles gambiae (from E. coli)
Catalog # PRSI90-359
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −80 °C.
  • Biological activity:
    Specific activity towards L-Trp: 87.0umol/hr per mg. TDO catalyzes the oxidative opening of the L-tryptophan indole moiety to yield N-formylkynurenine (NFK).
  • Protein synonyms:
    TRPO|Tryptophan Oxygenase|Tryptophan 2,3-dioxygenase|TPH2|Tryptophan Pyrrolase|Tryptophanase
  • Protein/peptide name:
    TDO Anopheles gambiae
  • Purity:
    >98% (SDS-PAGE)
  • Formulation:
    Liquid, In 50mM sodium phosphate pH 8.0, containing 0.15M sodium chloride, 0.2M imidazole and 10% glycerol.

Specifications

About this item

Tryptophan 2,3-dioxygenase (TDO) is a heme-containing dioxygenase catalyzing the addition of molecular oxygen across the 2,3-double bond of the indole ring of tryptophan to form N-formylkynurenine (NFK). In Anopheles gambiae, TDO is the only enzyme able to catalyze the first and rate-limiting step in L-Trp catabolism through the kynurenine pathway. Tryptophan is an essential amino acid for protein synthesis and also the precursor for production of a number of neurotransmitters, such as serotonin and melatonin; in mosquitoes, the kynurenine pathway is essential for eye pigmentation. Conceivably, the tryptophan-using pathways should be regulated in a coordinated manner in mosquitoes as well as in other species and TDO activation/inactivation processes could play an essential role in these phenomena.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.