Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant IL22 (from cells)
Human recombinant IL22 (from cells)
Catalog # PRSI92-661
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL22 (from cells)
Catalog # PRSI92-661
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-hIgG2 Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-TIF|IL-22|Cytokine Zcyto18|IL-10-related T-cell-derived-inducible factor|IL22|Interleukin-22
  • Protein/peptide name:
    IL22
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    43.4 kD
  • Sequence:
    Ala34-Ile179
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Interleukin-22(IL-22) is a member of a group of the IL-10 family, a class of potent mediators of cellular inflammatory responses. IL-22 is produced by activated DC and T cells. IL-22 and IL-10 receptor chains play a role in cellular targeting and signal transduction. It can initiate and regulate innate immune responses against bacterial pathogens especially in epithelial cells such as respiratory and gut epithelial cells. IL-22 along with IL-17 likely plays a role in the coordinated response of both adaptive and innate immune systems. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10. Biological activity of IL-22 is initiated by binding to a cell-surface complex consisting of IL-22R1 and IL-10R2 receptor chains. IL-22 biological activity is further regulated by interactions with a soluble binding protein, IL-22BP. IL-22BP and an extracellular region of IL-22R1 share sequence similarity. In some cases, the pro-inflammatory versus tissue-protective functions of IL-22 are regulated by cytokine IL-17A.

Fusion-Tag: C-hIgG2 Fc tag

This recombinant protein can be used for biological assays. For research use only.