Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant DDR2
Mouse recombinant DDR2
Catalog # PRSI92-640
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant DDR2
Catalog # PRSI92-640
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Receptor protein-tyrosine kinase TKT|receptor-related 3|CD167b|Neurotrophic tyrosine kinase|CD167 antigen-like family member B|Discoidin domain receptor 2|TKT|Discoidin domain-containing receptor 2|Tyrosine-protein kinase TYRO10|Ddr2
  • Protein/peptide name:
    DDR2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    43.5 kD
  • Sequence:
    Gln24-Arg399
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Discoidin domain receptor-2 (DDR2) is a cell surface tyrosine kinase receptor that can be activated by soluble collagen and has been implicated in diverse physiological functions including organism growth and wound repair. DDR2 binds to and is activated by collagen I, II, III, V, and X, with the notable exception of basement membrane collagen IV. DDR2 is expressed in connective tissues arising from embryonic mesoderm. DDR2 regulates cell proliferation, cell adhesion, migration, and extracellular matrix remodelling.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.