Human recombinant TIMP-4 (from cells)
Catalog # PRSI92-458
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Metalloproteinase inhibitor 4|TIMP-4|Tissue inhibitor of metalloproteinases 4|TIMP4
- Protein/peptide name:TIMP-4
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:23.5 kD
- Sequence:Cys30-Pro224
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Specifications
About this item
Metalloproteinase inhibitor 4 is an enzyme that in humans is encoded by the TIMP4 gene, belongs to the protease inhibitor I35 (TIMP) family. The protein complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Known to act on MMP-1, MMP-2, MMP-3, MMP-7 and MMP-9.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.