Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Calcitonin (from E. coli)
Human recombinant Calcitonin (from E. coli)
Catalog # PRSI92-181
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Calcitonin (from E. coli)
Catalog # PRSI92-181
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    PDN-21|CCP|CALC1|Calcitonin|Katacalcin|Calcitonin Carboxyl-Terminal Peptide|CALCA
  • Protein/peptide name:
    Calcitonin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    10.5 kD
  • Sequence:
    Tyr58-Asn141
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,50% Glycerol,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Calcitonin is a secreted protein which belongs to the calcitonin family. Calcitonin is cleaved into the following two chains: Calcitonin and Katacalcin. Katacalcin is a potent plasma calcium-lowering peptide. Calcitonin is a 32-amino acid linear polypeptide hormone. Calcitonin acts to reduce blood calcium (Ca2+), opposing the effects of parathyroid hormone (PTH). Its importance in humans has not been as well established as its importance in other animals, as its function is usually not significant in the regulation of normal calcium homeostasis. Calcitonin causes a rapid but short-lived drop in the level of calcium and phosphate in blood by promoting the incorporation of those ions in the bones.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.