Human recombinant CGREF1 (from cells)
Catalog # PRSI91-777
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Cell Growth Regulatory Gene 11 Protein|Cell Growth Regulator with EF Hand Domain Protein 1|Hydrophobestin|CGR11|CGREF1
- Protein/peptide name:CGREF1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:30.9 kD
- Sequence:Ala20-Ile301
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl,150mM NaCl,1mM GaCl2,pH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Cell Growth Regulator with EF Hand Domain Protein 1 (CGREF1) is a secreted calcium ion binding protein. CGREF1 contains two EF-hand domains and both EF-hands are required for function. Human CGREF1 is synthesized as a 301 amino acid precursor that contains a 19 amino acid signal sequence, and a 282 amino acid mature chain. CGREF1 is probably digested extracellularly by an unknown serine protease generating extremely hydrophobic bioactive peptides. CGREF1 mediates cell-cell adhesion in a calcium-dependent manner. In addition, CGREF1 is able to inhibit growth in several cell lines.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag