Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant proline-rich acidic 1 (from cells)
Human recombinant proline-rich acidic 1 (from cells)
Catalog # PRSI91-734
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant proline-rich acidic 1 (from cells)
Catalog # PRSI91-734
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Uterine-Specific Proline-Rich Acidic Protein|Epididymis Tissue Protein Li 178|Proline-Rich Acidic Protein 1|UPA|PRAP1
  • Protein/peptide name:
    Proline-Rich Acidic 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.04 kD
  • Sequence:
    Val21-Gln151
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Proline-rich acidic protein 1, also known as Uterine-specific proline-rich acidic protein, UPA and PRAP1, is a secreted protein. PRAP1 is abundantly expressed in the epithelial cells of the liver, kidney, gastrointestinal tract and cervix. PRAP1 is up-regulated by butyrate, trichostatin A and 5'-aza-2' deoxycytidine. PRAP1 may play an important role in maintaining normal growth homeostasis in epithelial cells. PRAP1 is suppressed through epigenetic mechanisms involving histone deacetylation and methylation. PRAP1 has been shown to cause cell growth inhibition in cancer cell lines.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag