Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant serpin F1 (from cells)
  PRSI91-488
 :  
Human recombinant serpin F1 (from cells)
  PRSI91-488
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    PEDF|Serpin F1|EPC-1|Pigment Epithelium-Derived Factor|SERPINF1|Cell Proliferation-Inducing Gene 35 Protein
  • Protein/peptide name:
    Serpin F1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    45.42 kD
  • Sequence:
    Gln20-Pro418
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Serpin F1 is a secreted glycoprotein that belongs to the noninhibitory serpin. It has an alpha/beta core serine-protease inhibitor domain, three major beta-sheets, and ten alpha-helices. As protease inhibitors, serpins have an array of functions including regulating blood clotting, the complement pathway, extracellular matrix remodeling, and cell motility. They are also involved in activities that extend beyond their ability to inhibit proteases. For instance, they may also regulate blood pressure, angiogenesis, or act as storage/transport proteins. Serpin F1 is a new promising approach for the treatment of osteosarcoma and has been described as a natural angiogenesis inhibitor with neurotrophic and immune-modulation properties. The human serpin superfamily consists of at least 35 members that target not only serine proteases, but also selected cysteine proteases and non-protease proteins. Levels of the natural ocular anti-angiogenic factor SentrinF1 (PEDF) is associated with proliferative retinopathy.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag