Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:MICA|PERB11.1|MIC-A|MHC Class I Polypeptide-Related Sequence A
- Protein/peptide name:MHC Class I-related A
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:33.87 kD
- Sequence:Glu24-Gln308
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:For Biological Assays
Specifications
About this item
MHC Class I Polypeptide-Related Sequence A (MICA) is a transmembrane glycoprotein that functions as a ligand for human NKG2D. Unlike classical MHC class I molecules, MICA does not form a heterodimer with beta-2-microglobulin. MICA shares 85% amino acid identity with a closely related protein, MICB. MICA acts as a stress-induced self-antigen that is recognized by NK cells, NKT cells, and most of the subtypes of T cells. As a Ligand for the KLRK1/NKG2D receptor, MICA binds to KLRK1 leads to cell lysis. MICA functions as an antigen for gamma delta T cells and is frequently expressed in epithelial tumors. MICA antigens are able to elicit the synthesis of alloantibodies in transplant recipients. Studies have shown that anti-MICA antibodies are associated with acute renal allograft rejection and failure. MICA recognition is involved in tumor surveillance, viral infections, and autoimmune diseases.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag